1,6-Hexanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-11-8
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00002985
:118.18 g/mol
Merck Index:13,04709
Boiling Pt:250 °C (1013 hPa)
Melting Pt:39…42 °C
Density:1.12 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:211-074-0