
1,4-Butanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-63-4
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00002968
:90.12 g/mol
Boiling Pt:230 °C (1013 hPa)
Melting Pt:19.5…20 °C
Density:1.017 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:203-786-5
:GHS07
Storage Temperature:Ambient