
2,3-Dimercapto-1-propanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:59-52-9
:
Formula:C₃H₈OS₂
MDL Number:MFCD00004864
Merck Index:13,03234
Boiling Pt:223 °C (1013 hPa)
Melting Pt:77 °C
Density:1.2531 g/cm³ (20 °C)
Flash Pt:112 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:200-433-7
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:124.22 g/mol
:GHS07