Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethanol denatured
Ethanol denatured ≥96% (denatured with 1% MEK, 1% IPA and 10 ppm Bitrex®)
Ethanol denatured ≥96% (denatured with 1% MEK, 1% IPA and 10 ppm Bitrex®)
:
Roth Carl
Ethanol denatured ≥96% (denatured with 1% MEK, 1% IPA and 10 ppm Bitrex®)
:
Roth Carl
:
64-17-5
:
:
H₃CCH₂OH
:
MFCD00003568
:
46.07 g/mol
:
–114 °C
:
Ambient
:
200-578-6
:
1170
:
3,II
:
GHS02