
1-Hexanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-27-3
:
Formula:H₃C(CH₂)₄CH₂OH
MDL Number:MFCD00002982
:102.18 g/mol
Merck Index:13,04715
Boiling Pt:156 °C (1013 hPa)
Melting Pt:–45 °C
Density:0.8322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-852-3
UN:2282
ADR:3,III
:M-Clarity™ quality level = MQ300
Flash Pt:60 °C
:GHS07