1-Heptanol
Glass bottle, 500 mL
Catalog # ROTH3827.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Heptyl alcohol
- CAS Number:111-70-6
- Formula:H₃C(CH₂)₅CH₂OH
- MDL Number:MFCD00002986
- Molecular Weight:116.2 g/mol
- Merck Index:13,04681
- Boiling Pt:176 °C (1013 hPa)
- Melting Pt:–35 °C
- Density:0.82 g/cm³ (20 °C)
- Flash Pt:70 °C
- Storage Temperature:Ambient
- EINECS:203-897-9
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL



