Glutaric anhydride 95%
:108-55-4
:
:C₅H₆O₃
:MFCD00006679
:114.1 g/mol
:290 °C (760 mmHg)
:46…57 °C
:1,411 g/cm³ (20 °C)
:173 °C
:Ambient
:203-593-6
:GHS05
:GHS07
- Appearance:White to yellow to light beige Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- GC:≥94 %
- HPLC:≥97.5 %
- NMR (1H-NMR):Consistent with structure