Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Nitroethanol tech. 80%, Technical Grade

Irritant
2-Nitroethanol tech. 80%, Technical Grade
Certificates
2-Nitroethanol tech. 80%, Technical Grade
2-Nitroethanol tech. 80%, Technical Grade

3 Options Available Below

Product Details & Documents

Purity: 80% CAS No.: 625-48-9 Molecular Formula: O2NCH2CH2OH Formula Weight: 91.07 Flash Point: 110°(230°F) Refractive Index: 1.4451 Melting Point: -80° Boiling Point: 194° Beilstein Reference: 1633753 MDL No.: MFCD00007405

  • CAS Number:625-48-9
  • Formula:C₂H₅NO₃
  • MDL Number:MFCD00007405
  • Molecular Weight:91.07 g/mol
  • Boiling Pt:194 °C
  • Melting Pt:–80 °C
  • Density:1.270
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:210-895-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 56,50 €
    56,50 €
    1 * 5 g
       56,50 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 186,00 €
    186,00 €
    1 * 25 g
       186,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 504,00 €
    504,00 €
    1 * 100 g
       504,00 €