Order Entry
Export
ContactUsLinkComponent
1,8-Diaminooctane 98%
Corrosive
Irritant
1,8-Diaminooctane 98%
Certificates
undefined
Corrosive
Irritant
1,8-Diaminooctane 98%

Specification Test Results

  • Appearance:
    White to yellow Fused crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    50 to 54 °C
  • GC:
    ≥97.5 %
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information