Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,7-Diaminoheptane
1,7-Diaminoheptane, Sigma-Aldrich®
1,7-Diaminoheptane, Sigma-Aldrich®
:
Merck
1,7-Diaminoheptane, Sigma-Aldrich®
:
Merck
:
646-19-5
:
:
C₇H₁₈N₂
:
MFCD00008246
:
130.23 g/mol
:
224 °C (1013 hPa)
:
28…29 °C
:
1.012 g/cm³ (20 °C)
:
211-468-2
:
3,II
:
M-Clarity™ quality level = MQ100
:
3295
:
GHS05
:
Refrigerator