

1,7-Diaminoheptane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:646-19-5
:
Formula:C₇H₁₈N₂
MDL Number:MFCD00008246
:130.23 g/mol
Boiling Pt:224 °C (1013 hPa)
Melting Pt:26…30 °C
Density:1,012 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Refrigerator
EINECS:211-468-2
UN:3259
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to white to yellow Liquid or solid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with HClO4:97.5 to 102.5 %