

1,6-Diaminohexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:124-09-4
:
Formula:C₆H₁₆N₂
MDL Number:MFCD00008243
:116.21 g/mol
Merck Index:13,04713
Boiling Pt:205 °C (1013 hPa)
Melting Pt:39…42 °C
Density:0.66 g/cm³ (20 °C)
EINECS:204-679-6
UN:2280
ADR:8,III
:M-Clarity™ quality level = MQ200
Flash Pt:80 °C
:GHS05
:GHS07
Storage Temperature:Ambient