Order Entry
Export
ContactUsLinkComponent
1,6-Diaminohexane
Corrosive
Irritant
1,6-Diaminohexane
Catalog # ROTH4465.1
Supplier:  Roth Carl
CAS Number:  
Corrosive
Irritant
1,6-Diaminohexane
Catalog # ROTH4465.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :116.21 g/mol
Merck Index:13,04713
Boiling Pt:205 °C (1013 hPa)
Melting Pt:39…42 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:80 °C
Storage Temperature:Ambient
EINECS:204-679-6
UN:2280
ADR:8,III
 :GHS05
 :GHS07
CAS Number:124-09-4
Formula:C₆H₁₆N₂
MDL Number:MFCD00008243