Order Entry
Export
ContactUsLinkComponent
1,4-Diaminobutane
1,4-Diaminobutane
Catalog # EHERC12193000
Supplier:  EHRENSTORFER
CAS Number:  
1,4-Diaminobutane
Catalog # EHERC12193000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :88.15 g/mol
Merck Index:12,08134
Boiling Pt:158 °C (1013 hPa)
Melting Pt:25…28 °C
Density:0.88 g/cm³ (20 °C)
Flash Pt:63 °C
Storage Temperature:Ambient
EINECS:203-782-3
UN:2928
ADR:6.1,II
CAS Number:110-60-1
Formula:C₄H₁₂N₂
MDL Number:MFCD00008235