Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diaminopropane 99%

1,3-Diaminopropane 99%

Flammable
Corrosive
Toxic
Health
1,3-Diaminopropane 99%
Certificates
1,3-Diaminopropane 99%

3 Options Available Below

Product Details & Documents
Synonyms:  1,3-Propanediamine, Trimethylenediamine
  • CAS Number:109-76-2
  • Formula:C₃H₁₀N₂
  • MDL Number:MFCD00008228
  • Molecular Weight:74.13 g/mol
  • Boiling Pt:140 °C (760 mmHg)
  • Melting Pt:–12 °C
  • Density:0.88 g/cm³
  • Flash Pt:48 °C
  • Storage Temperature:Ambient
  • EINECS:203-702-7
  • UN:2734
  • ADR:8,I

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4565 to 1.4585 (20°C, 589 nm)
  • Water:
    ≤0.5 %
  • GC:
    ≥98.5 %
  • Color scale:
    ≤20 APHA
  • Titration with HClO4:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 24,00 €
    24,00 €
    1 * 50 mL
       24,00 €

  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 60,70 €
    60,70 €
    1 * 250 mL
       60,70 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 167,00 €
    167,00 €
    1 * 1 L
       167,00 €