



1,2-Propylenediamine 99%
:78-90-0
:
:C₃H₁₀N₂
:MFCD00008089
:74.13 g/mol
:12,08037
:119…120 °C (760 mmHg)
:–37 °C
:0.87 g/cm³
:33 °C
:201-155-9
:2258
:8,II
:GHS06
:GHS05
:GHS02
:GHS08
:Ambient
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4470 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥98.5 %
- Color scale:≤20 APHA