ContactUsLinkComponent
Taurine, SAFC®
Taurine, SAFC®
  SIALT4571-1KG
 :  Merck
 :  
Taurine, SAFC®
  SIALT4571-1KG
 :  Merck
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024
 :
 :125.15 g/mol
 :13,09163
 :300 °C
 :M-Clarity™ quality level = MQ500 (more info.)
 :Ambient
 :203-483-8
 :107-35-7
 :C₂H₇NO₃S
 :MFCD00008197