Order Entry
Export
Orders LinkContactUsLinkComponent
 

Taurine

Glass bottle for solids, 1 kg
Catalog # MOLE13830142-1KG
Supplier:   Molekula
Taurine
Catalog # MOLE13830142-1KG
Supplier:  Molekula
CAS Number:  107-35-7
Taurine

Glass bottle for solids, 1 kg

CAS Number:  107-35-7
Price
205,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  2-Aminoethanesulphonic acid
  • CAS Number:107-35-7
  • Formula:C₂H₇NO₃S
  • MDL Number:MFCD00008197
  • Molecular Weight:125.15 g/mol
  • Merck Index:13,09163
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:203-483-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications