Taurine 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 2-Aminoethanesulphonic acid
- CAS Number:107-35-7
- Formula:C₂H₇NO₃S
- MDL Number:MFCD00008197
- Molecular Weight:125.15 g/mol
- Merck Index:13,09163
- Melting Pt:300 °C (dec.)
- Storage Temperature:Ambient
- EINECS:203-483-8
4 Options Available Below
Product Details & Documents
Synonyms: 2-Aminoethanesulphonic acid
- CAS Number:107-35-7
- Formula:C₂H₇NO₃S
- MDL Number:MFCD00008197
- Molecular Weight:125.15 g/mol
- Merck Index:13,09163
- Melting Pt:300 °C (dec.)
- Storage Temperature:Ambient
- EINECS:203-483-8
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 %
- Sulfated ash:≤0.1 %
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤110 ppm
- Sulfate (SO4):≤140 ppm
- Ammonium (NH4):≤200 ppm
- Arsenic (As):≤2 ppm
- Heavy metals (as Pb):≤20 ppm



