0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
:
Roth Carl
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
:
Roth Carl
:
77-86-1
:
:
NH₂C(CH₂OH)₃
:
MFCD00004679
:
121.14 g/mol
:
13,09842
:
357 °C (1013 hPa)
:
169…172 °C
:
1.328 g/cm³ (20 °C)
:
201-064-4
:
GHS07
:
Ambient