Safety shoes, lace-up, jo_Twist Boa Low, Jori
Supplier: ELTEN
Certificates
About this item
Antistatic safety shoe made from hydrophobised nubuck leather and hydrophobised textile material, a full, padded bellows tongue, and a dual-density PU/PU sole (oil, fuel and slip resistant, heat resistant up to approx. 130 °C).
- Climate regulating, breathable textile lining
- Lace- and buckle-free Boa® system can be opened and closed with just one hand
- Full-length inlay sole
- Protective composite toecap (200 J)
- Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN ISO 20345; S1P SRC
Specifications
- Catalog No:
- ELTN12761/47
- ELTN12761/45
- ELTN12761/46
- ELTN12761/42
- ELTN12761/43
- ELTN12761/41
- ELTN12761/40
- ELTN12761/44
- ELTN12761/39
- ELTN12761/38
- ELTN12761/48
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001