Ethanolamine
Supplier: Roth Carl
Jump to:
About this item
Synonyms: 2-Aminoethanol, Monoethanolamine
- CAS Number:141-43-5
- Formula:NH₂CH₂CH₂OH
- MDL Number:MFCD00008183
- Molecular Weight:61.08 g/mol
- Merck Index:13,03762
- Boiling Pt:171 °C (1013 hPa)
- Melting Pt:10.5 °C
- Density:1.0164 g/cm³ (20 °C)
- Flash Pt:93 °C
- Storage Temperature:Ambient
- EINECS:205-483-3
- UN:2491
- ADR:8,III
2 Options Available Below
Product Details & Documents
Synonyms: 2-Aminoethanol, Monoethanolamine
- CAS Number:141-43-5
- Formula:NH₂CH₂CH₂OH
- MDL Number:MFCD00008183
- Molecular Weight:61.08 g/mol
- Merck Index:13,03762
- Boiling Pt:171 °C (1013 hPa)
- Melting Pt:10.5 °C
- Density:1.0164 g/cm³ (20 °C)
- Flash Pt:93 °C
- Storage Temperature:Ambient
- EINECS:205-483-3
- UN:2491
- ADR:8,III



