Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iodomethane
Iodomethane, Sigma-Aldrich®
Iodomethane, Sigma-Aldrich®
:
Merck
Iodomethane, Sigma-Aldrich®
:
Merck
:
74-88-4
:
:
CH₃I
:
MFCD00001073
:
141.94 g/mol
:
12,06161
:
42.4 °C (1013 hPa)
:
–66 °C
:
2.279 g/cm³ (20 °C)
:
2644
:
6.1,I
:
M-Clarity™ quality level = MQ200
:
200-819-5
:
GHS06
:
GHS08
:
Refrigerator