
1,5-Diiodopentane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:628-77-3
:
Formula:C₅H₁₀I₂
MDL Number:MFCD00001101
:323.94 g/mol
Boiling Pt:264 °C (1013 hPa)
Melting Pt:9 °C
Density:2.195 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:211-054-1
:GHS07