Trimethylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:27607-77-8
:
Formula:C₄H₉F₃O₃SSi
MDL Number:MFCD00000406
:222.26 g/mol
Merck Index:12,09849
Boiling Pt:77 °C (1013 hPa)
Density:1.225 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:248-565-4