1,4-Diiodoperfluorobutane, Sigma-Aldrich®
Catalog # SIAL473863-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:453.84 g/mol
Boiling Pt:150 °C (1013 hPa)
Melting Pt:–3 °C
Density:2.5466 g/cm³ (0 °C)
:M-Clarity™ quality level = MQ100
EINECS:206-788-4
CAS Number:375-50-8
Formula:C₄F₈I₂
MDL Number:MFCD00042263