

Thioacetamide ≥99.0% analytical reagent
28603.137
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Assay:Min. 99.0 %
- Melting point:110 - 113 °C
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 0.05 %
- Fe (Iron):Max. 5 ppm
:
:75.13 g/mol
:13,09391
:113…114 °C
:1.336 g/cm³ (20 °C)
:200-541-4
:GHS08
:GHS07
:62-55-5
:CH₃CSNH₂
:MFCD00008070