Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium citrate monohydrate, Sigma-Aldrich®

Potassium citrate monohydrate, Sigma-Aldrich®

Glass bottle for solids, 500 g
Catalog # SIALC3029-500G
Supplier:   Merck
Potassium citrate monohydrate, Sigma-Aldrich®
Catalog # SIALC3029-500G
Supplier:  Merck
CAS Number:  6100-05-6
Potassium citrate monohydrate, Sigma-Aldrich®
Potassium citrate monohydrate, Sigma-Aldrich®

Glass bottle for solids, 500 g

CAS Number:  6100-05-6
Price
60,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate
  • CAS Number:6100-05-6
  • Formula:HOC(COOK)(CH₂COOK)₂·H₂O
  • MDL Number:MFCD00150442
  • Molecular Weight:324.41 g/mol
  • Merck Index:13,07706
  • Melting Pt:230 °C
  • Density:1.98 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • EINECS:212-755-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications