Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium citrate monohydrate

Potassium citrate monohydrate

Glass bottle for solids, 1 kg
Catalog # MOLE21508672-1KG
Supplier:   Molekula
Potassium citrate monohydrate
Catalog # MOLE21508672-1KG
Supplier:  Molekula
CAS Number:  6100-05-6
Potassium citrate monohydrate
Potassium citrate monohydrate

Glass bottle for solids, 1 kg

CAS Number:  6100-05-6
Price
99,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate
  • CAS Number:6100-05-6
  • Formula:HOC(COOK)(CH₂COOK)₂·H₂O
  • MDL Number:MFCD00150442
  • Molecular Weight:324.41 g/mol
  • Merck Index:13,07706
  • Melting Pt:230 °C
  • Density:1.98 g/cm³ (20 °C)
  • EINECS:212-755-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications