Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
di-Potassium oxalate monohydrate
di-Potassium oxalate monohydrate
di-Potassium oxalate monohydrate
:
Roth Carl
di-Potassium oxalate monohydrate
:
Roth Carl
:
6487-48-5
:
:
(COOK)₂·H₂O
:
MFCD00150033
:
184.23 g/mol
:
12,07820
:
356 °C
:
2.154 g/cm³ (20 °C)
:
Ambient
:
209-506-8
:
2811
:
6.1,III
:
GHS07