Order Entry
Export
Orders LinkContactUsLinkComponent
1,12-Dodecanedioic acid, Sigma-Aldrich®

1,12-Dodecanedioic acid, Sigma-Aldrich®

Irritant
Supplier:   Merck
1,12-Dodecanedioic acid, Sigma-Aldrich®
Supplier:  Merck
Certificates
1,12-Dodecanedioic acid, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Dodecanedioic acid, 1,10-Decanedicarboxylic acid
  • CAS Number:693-23-2
  • Formula:C₁₂H₂₂O₄
  • MDL Number:MFCD00002735
  • Molecular Weight:230.3 g/mol
  • Boiling Pt:294 °C (1013 hPa)
  • Melting Pt:121…126 °C
  • Density:1.175 g/cm³ (21 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:220 °C
  • Storage Temperature:Freezer
  • EINECS:211-746-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 52,30 €
    52,30 €
    1 * 100 g
       52,30 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 79,60 €
    79,60 €
    1 * 500 g
       79,60 €