Order Entry
Export
Orders LinkContactUsLinkComponent
1,12-Dodecanedioic acid ≥99%

1,12-Dodecanedioic acid ≥99%

Irritant
   Sustainable Options Available
1,12-Dodecanedioic acid ≥99%
   Sustainable Options Available
Certificates
1,12-Dodecanedioic acid ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 693-23-2 Molecular Formula: HO2C(CH2)10CO2H Formula Weight: 230.31 Flash Point: 227°(440°F) Odour: Odourless Boiling Point: 244-245°/10mm Beilstein Reference: 1782580 MDL No.: MFCD00002735

Synonyms:  Dodecanedioic acid, 1,10-Decanedicarboxylic acid
  • CAS Number:693-23-2
  • Formula:C₁₂H₂₂O₄
  • MDL Number:MFCD00002735
  • Molecular Weight:230.3 g/mol
  • Boiling Pt:244…245 °C (10 mmHg)
  • Melting Pt:127…131 °C
  • Density:1.150
  • Flash Pt:227 °C (440 °F)
  • Storage Temperature:Ambient
  • EINECS:211-746-3
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 46,90 €
    46,90 €
    1 * 250 g
       46,90 €

    Specifications:
    More
    Specifications
    Cat. No.
    A10387.30
  • Glass bottle for solids
    1.000 g
    Log in for availability
    Log in for availability
    1 * 1.000 g
    1 * 1.000 g - 204,00 €
    204,00 €
    1 * 1.000 g
       204,00 €

    Specifications:
      
    More
    Specifications
    Cat. No.
    A10387.0B