Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,12-Dodecanedioic acid 99%

Irritant
1,12-Dodecanedioic acid 99%
Certificates
1,12-Dodecanedioic acid 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Dodecanedioic acid, 1,10-Decanedicarboxylic acid
  • CAS Number:693-23-2
  • Formula:C₁₂H₂₂O₄
  • MDL Number:MFCD00002735
  • Molecular Weight:230.3 g/mol
  • Boiling Pt:245 °C (10 mmHg)
  • Melting Pt:125…130 °C
  • Density:1,175 g/cm³ (21 °C)
  • Flash Pt:220 °C
  • Storage Temperature:Ambient
  • EINECS:211-746-3

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder or pellets
  • Infrared spectrum:
    Conforms
  • Melting point:
    125 to 130 °C
  • Titration with NaOH:
    ≥98.5 %
  • Viscosity:
    30.5 to 36.5
  • pH:
    3.0 to 3.8 (1 % solution)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,70 €
    21,70 €
    1 * 5 g
       21,70 €

  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 27,40 €
    27,40 €
    1 * 100 g
       27,40 €

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 67,80 €
    67,80 €
    1 * 500 g
       67,80 €