Sarcosine
Glass bottle for solids, 1 kg
Catalog # MOLEM29103605
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
- CAS Number:107-97-1
- Formula:C₃H₇NO₂
- MDL Number:MFCD00004279
- Molecular Weight:89.09 g/mol
- Merck Index:12,08519
- Boiling Pt:185 °C (1013 hPa)
- Melting Pt:63 °C
- Density:1.301 g/cm³ (20 °C)
- EINECS:203-538-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



