Order Entry
Export
Orders LinkContactUsLinkComponent
Sarcosine

Sarcosine

Sarcosine
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
  • CAS Number:107-97-1
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00004279
  • Molecular Weight:89.09 g/mol
  • Merck Index:12,08519
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:63 °C
  • Density:1.301 g/cm³ (20 °C)
  • EINECS:203-538-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g