Order Entry
Export
Orders LinkContactUsLinkComponent
Sarcosine ≥98%

Sarcosine ≥98%

Sarcosine ≥98%
Certificates
Sarcosine ≥98%
Sarcosine ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 107-97-1 Molecular Formula: CH3NHCH2CO2H Formula Weight: 89.09 Odour: Odourless pH: 6.1 Melting Point: ca 208° dec. Beilstein Reference: 1699442 Merck Reference: 14,8373 MDL No.: MFCD00004279

Synonyms:  H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
  • CAS Number:107-97-1
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00004279
  • Molecular Weight:89.09 g/mol
  • Merck Index:14,8373
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:∼ 208 °C (dec.)
  • Density:1.301 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-538-6
Product Family Options

Product Information