Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Aminocaproic acid
6-Aminocaproic acid
6-Aminocaproic acid
:
Roth Carl
6-Aminocaproic acid
:
Roth Carl
:
60-32-2
:
:
C₆H₁₃NO₂
:
MFCD00008238
:
131.17 g/mol
:
13,00431
:
257 °C (1013 hPa)
:
127 °C
:
207 °C
:
200-469-3
:
GHS07
:
Ambient