Order Entry
Export
ContactUsLinkComponent
6-Aminocaproic acid
6-Aminocaproic acid
Catalog # MOLE15845021-1KG
Supplier:  Molekula
CAS Number:  
6-Aminocaproic acid
Catalog # MOLE15845021-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :131.17 g/mol
Merck Index:13,00431
Boiling Pt:257 °C (1013 hPa)
Melting Pt:127 °C
Flash Pt:207 °C
Storage Temperature:Ambient
EINECS:200-469-3
 :GHS07
CAS Number:60-32-2
Formula:C₆H₁₃NO₂
MDL Number:MFCD00008238