
Manganese(II) acetate, anhydrous 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:638-38-0
:
Formula:(H₃CCOO)₂Mn
MDL Number:MFCD00013039
:173.03 g/mol
Merck Index:14,5724
EINECS:211-334-3
:GHS07
Storage Temperature:Ambient