Order Entry
Export
Orders LinkContactUsLinkComponent
Zinc stearate

Zinc stearate

Supplier:   Roth Carl
Zinc stearate
Supplier:  Roth Carl
Certificates
Zinc stearate

2 Options Available Below

Product Details & Documents
Synonyms:  Stearic acid zinc salt, Zinc(II) stearate
  • CAS Number:557-05-1
  • Formula:C₃₆H₇₀O₄Zn
  • MDL Number:MFCD00013031
  • Molecular Weight:632.34 g/mol
  • Merck Index:13,10212
  • Melting Pt:140 °C
  • Density:1.095 g/cm³ (20 °C)
  • Flash Pt:276 °C
  • Storage Temperature:Ambient
  • EINECS:209-151-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 75,30 €
    75,30 €
    1 * 1 kg
       75,30 €

  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 913,00 €
    913,00 €
    1 * 25 kg
       913,00 €