Zinc stearate
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Stearic acid zinc salt, Zinc(II) stearate
- CAS Number:557-05-1
- Formula:C₃₆H₇₀O₄Zn
- MDL Number:MFCD00013031
- Molecular Weight:632.34 g/mol
- Merck Index:13,10212
- Melting Pt:118…128 °C
- Density:1,095 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:209-151-9
3 Options Available Below
Product Details & Documents
Synonyms: Stearic acid zinc salt, Zinc(II) stearate
- CAS Number:557-05-1
- Formula:C₃₆H₇₀O₄Zn
- MDL Number:MFCD00013031
- Molecular Weight:632.34 g/mol
- Merck Index:13,10212
- Melting Pt:118…128 °C
- Density:1,095 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:209-151-9
Documents
ACRO26378
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration Complexometric:12.5 to 14.0 % (ZnO)



