


Mercury(II) acetate ≥98.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 25364.186
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:Min. 98.5 %
- Substances not reducing by formic acid:Max. 0.1 %
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 50 ppm
- Fe (Iron):Max. 10 ppm
- Hg (I) (Mercury):Max. 0.3 %
Specification Test Results
About this item
:
:318.68 g/mol
Merck Index:13,05898
Melting Pt:178…180 °C
Density:3.27 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-491-1
UN:1629
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:1600-27-7
Formula:(H₃CCOO)₂Hg
MDL Number:MFCD00012165