


Mercury(II) acetate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1600-27-7
:
Formula:(H₃CCOO)₂Hg
MDL Number:MFCD00012165
:318.68 g/mol
Merck Index:14,5873
Melting Pt:179…182 °C
Density:3.28
EINECS:216-491-1
UN:1629
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient