

Copper(II) acetate, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:142-71-2
:
Formula:CuCH₃CO₂
MDL Number:MFCD00008690
:181.64 g/mol
Merck Index:14,2624
Boiling Pt:240 °C (1013 hPa)
Melting Pt:115 °C
Density:1.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-553-3
UN:3077
ADR:9,III
:GHS05
:GHS07
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organometallic