
Ammonium formate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:540-69-2
:
Formula:HCO₂NH₄
MDL Number:MFCD00013103
:63.06 g/mol
Merck Index:13,00525
Melting Pt:119…121 °C
Density:1.28 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
EINECS:208-753-9
:GHS07
Storage Temperature:Ambient