Ammonium acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Acetic acid ammonium salt
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
:77.08 g/mol
Merck Index:13,00495
Melting Pt:114 °C
Density:1.17 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:136 °C
EINECS:211-162-9
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: