Order Entry
Export
Orders LinkContactUsLinkComponent
Ammonium acetate, Sigma-Aldrich®

Ammonium acetate, Sigma-Aldrich®

Irritant
Supplier:   Merck
Ammonium acetate, Sigma-Aldrich®
Supplier:  Merck
Certificates
Ammonium acetate, Sigma-Aldrich®
Ammonium acetate, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid ammonium salt
  • CAS Number:631-61-8
  • Formula:H₃CCOONH₄
  • MDL Number:MFCD00013066
  • Molecular Weight:77.08 g/mol
  • Merck Index:13,00495
  • Melting Pt:114 °C
  • Density:1.17 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Flash Pt:136 °C
  • Storage Temperature:Ambient
  • EINECS:211-162-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 89,40 €
    89,40 €
    1 * 10 g
       89,40 €

  • Plastic bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 446,00 €
    446,00 €
    1 * 100 g
       446,00 €