Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ammonium acetate

Irritant
Supplier:   Roth Carl
Ammonium acetate
Supplier:  Roth Carl
Certificates
Ammonium acetate
Ammonium acetate

4 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid ammonium salt
  • CAS Number:631-61-8
  • Formula:H₃CCOONH₄
  • MDL Number:MFCD00013066
  • Molecular Weight:77.08 g/mol
  • Merck Index:13,00495
  • Melting Pt:114 °C
  • Density:1.17 g/cm³ (20 °C)
  • Flash Pt:136 °C
  • Storage Temperature:Ambient
  • EINECS:211-162-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 36,90 €
    36,90 €
    1 * 500 g
       36,90 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 58,70 €
    58,70 €
    1 * 1 kg
       58,70 €

  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 101,00 €
    101,00 €
    1 * 2,5 kg
       101,00 €

  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 691,00 €
    691,00 €
    1 * 25 kg
       691,00 €