Sodium formate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:141-53-7
:
Formula:HCO₂Na
MDL Number:MFCD00013101
:68.01 g/mol
Merck Index:13,08694
Melting Pt:257.31 °C
Density:1.919 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:205-488-0
REACH:01-2119486468-21