Sodium acetate
Catalog # MOLE49106710-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:82.03 g/mol
Merck Index:12,08711
Boiling Pt:881.4 °C (1013 hPa)
Melting Pt:324 °C
Density:1.528 g/cm³ (20 °C)
Flash Pt:>250 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459